Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Mouse Anti-Human Chromogranin A, BD Horizon™
Mouse Monoclonal Antibody
$188.00 - $468.00
Specifications
Antigen | Chromogranin A |
---|---|
Clone | S21-537 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
Chromogranin A | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I | |
Human Chromogranin A Recombinant Protein | |
Affinity Chromatography |
S21-537 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
1113 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title