Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Hamster Anti-Mouse Notch4, BD Horizon™
Hamster Monoclonal Antibody
$122.00 - $306.00
Specifications
Antigen | Notch4 |
---|---|
Clone | HMN4-14 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Notch4 Monoclonal antibody specifically detects Notch4 in Mouse samples. It is validated for Flow cytometrySpecifications
Notch4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
18132 | |
IgG κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HMN4-14 | |
Flow Cytometry | |
R718 | |
Hamster | |
Mouse | |
Notch 4; Notch gene homolog 4; Notch4 | |
Mouse Extracellular Notch4 Domain Recombinant Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title