Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Edaglitazone
Potent and selective PPARγ agonist; antidiabetic
$233.50 - $979.50
Specifications
Chemical Name or Material | Edaglitazone |
---|---|
CAS | 213411-83-7 |
Synonym | 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione |
Solubility | Soluble to 100mM in DMSO |
Physical Form | Solid |
Description
Potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.Specifications
Edaglitazone | |
5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione | |
Solid | |
>98% |
213411-83-7 | |
Soluble to 100mM in DMSO | |
Bioactive Small Molecule |
Safety and Handling
Recommended Storage : Store at 4°C
Research Use Only. Not For Human or Veterinary Use
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title