Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid, 98%, Thermo Scientific™

Catalog No. AAH5200303
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 500789-06-0
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD03427959
InChI Key UDUKZORPLJUWTF-LLVKDONJSA-N
Synonym r-3-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, boc-r-3-amino-3-3-chlorophenyl propionic acid, r-3-boc-amino-3-3-chlorophenyl propionic acid, 3r-3-tert-butoxy carbonylamino-3-3-chlorophenyl propanoic acid, 3r-3-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, boc-d-?-phe 3-cl-oh, boc-d-beta-phe 3-cl-oh, boc-phg 3-cl-c*ch2 oh, r-boc-3-chloro-beta-phenylalanine, boc-d-3-amino-3-3-chlorophenyl-propionic acid
PubChem CID 2761720
IUPAC Name (3R)-3-(3-chlorophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)Cl

Specifications

Melting Point 90°C to 92°C
Quantity 1 g
Optical Rotation +40° (c=1 in Ethanol)
Formula Weight 299.75
Percent Purity 98%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.