Learn More
Hach Company 1cm x 1 cm Pour Through Cell, 450µL in Quartz glass
Supplier:  Hach Company LZV510
Encompass_Preferred
Four pour thru application in combination with Sipper modules.
Catalog No. 50-199-7909
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.