Learn More
Bioassay Systems QUANTICHROM GLUCOSE ASSAY KIT
Supplier: Bioassay Systems DIGL100
Assay kit, glucose and evaluation of drug effects on glucose metabolism. Method: OD630nm (Chemical). Samples: biological, food and beverage. Species: all. Procedure: 10 min. Size: 100 tests. Detection limit: 0.7 mg/dL (39 uM).