Learn More
Bioassay Systems QuantiChrom&trade Chromium Assay Kit. For quantitative determination of chromium Cr(VI).
SDP
Supplier:  Bioassay Systems DCRM250
Encompass_Preferred
For quantitative determination of chromium Cr(VI). Key Features: Sensitive and accurate. Linear detection range of 20 - 2000 µg/L Chromium. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated for processing thousands of samples per day. Method: OD480nm. Samples: Biological (serum, plasma etc), environmental (water, soil etc), food and beverage samples. Species: All. Procedure: Assay takes 20 min. Kit size: 250 tests. Detection limit: 20 µg/L (0.38 µM, 0.02 ppm).
Catalog No. 50-489-203
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.