Learn More
CTL Scientific Supply Corp QUANT. PERACETIC ACID 100 STR
Supplier: CTL Scientific Supply Corp 91342
QUANTOFIX PERACETIC ACID 2000 are test sticks for the semi-quantitative determination of peracetic acid. Gradation 500 - 1000 - 1500 - 2000 mg/l. Contains 100 test strips