Learn More
Neuromics, Inc Heat Inactivated Qualified Imported Fetal Bovine Serum (USDA Approved), 500 ml is ideal for the general cell culturing applications. It is triple 0.1 um filtered and free of virus and mycoplasma.
Supplier:  Neuromics, Inc FBS006HI
Encompass
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Heat Inactivated Qualified Imported (USDA Approved) FBS features: Triple 0.1 μm Filtered and free of Virus and Mycoplasma.This treatment allows to destroy the complement acting in the immune system to avoid an interference with some experimentation, in particular the tests of immunology. Excellent for research involving immune response, viral studies, cytotoxicity assays, and stem cells.
Catalog No. NC2071937
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.