Learn More
Enzo Life Sciences AMPIGENE qPCR 1-Step Probe Kit Lo-ROX (1000Reactions)
SDP
Supplier:  Enzo Life Sciences ENZNUC1101000
Encompass_Preferred
AMPIGENE qPCR 1-Step Probe Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE qPCR 1-Step Probe Kits are designed for use on a variety of probes including: Taqman, Molecular Beacons, and Scorpion probes. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE qPCR 1-Step Probe Kits come with either Lo-ROX, Hi-ROX, or No-ROX concentration. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.) Applications: PCR.
Catalog No. 50-201-3888
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.