Learn More
Enzo Life Sciences AMPIGENE qPCR 1-Step Green Kit Lo-ROX (200Reactions)
Supplier: Enzo Life Sciences ENZNUC1080200
AMPIGENE qPCR 1-Step Green Kits use the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and qPCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. AMPIGENE qPCR 1-Step Green Kits use a proprietary intercalating dye that does not inhibit PCR. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE qPCR 1-Step Green Kits come with either Lo-ROX or Hi-ROX concentration. For Hi-ROX, see ENZ-NUC109. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.) Applications: PCR.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.