Learn More
Tecan 96-Well Verification Plates
Supplier: Tecan 30062826
For verification of liquid handling performance for QC Kit. 25 plates/sales unit. The Tecan QC Kit is a quality control tool for pipetting volume verification of your liquid handling platform, ensuring full regulatory compliance.
This item is not returnable.
View return policy