Learn More
New England Biolabs, Inc. Q5 High-Fidelity PCR Kit – 50 reactions
Supplier: New England Biolabs, Inc. E0555S
Q5 High-Fidelity DNA Polymerase sets a new standard for both fidelity and robust performance. With the highest fidelity amplification available (~280 times higher than Taq), Q5 DNA Polymerase results in ultra-low error rates. Q5 DNA Polymerase is composed of a novel polymerase that is fused to the processivity-enhancing Sso7d DNA binding domain, improving speed, fidelity and reliability of performance. This kit contains a sufficient supply of master mix and nuclease-free water to perform 50 polymerase chain reactions or 200 reactions. For added convenience, gel loading dye, and a Quick-Load DNA LAdder are also included in this kit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.