Learn More
Bioassay Systems EnzyChrom™ Pyruvate Kinase Assay Kit. For quantitative determination of pyruvate kinase activity and evaluation of drug effects on PK activity.
SDP
Supplier:  Bioassay Systems EPRK100
Encompass_Preferred
For quantitative determination of pyruvate kinase (PK) activity and evaluation of drug effects on PK activity. Key Features: Sensitive and accurate. Linear detection range in 96-well plate: 0.1 to 50 U/L for colorimetric assays and 0.01 to 2 U/L for fluorimetric assays run at 25°C for 30 min. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, FL530/590nm. Samples: Plasma, serum, cell and tissue etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.01 U/L.
Catalog No. 50-489-289
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.