Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purus™ PurusMat Frames: PROMO

Designed for use with Purus Mats and Purus IntroMats

Specifications

Color White
For Use With (Application) Cleanrooms
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-125-101PM
View Documents
Purus™
PF 2047 01 W 00PM
20 x 47 in. N/A N/A N/A
19-125-102PM
View Documents
Purus™
PF 2638 01 W 00PM
26 x 38 in. N/A N/A N/A
19-125-103PM
View Documents
Purus™
PF 2847 01 W 00PM
28 x 47 in. N/A N/A N/A
19-125-105PM
View Documents
Purus™
PF 3847 01 W 00PM
38 x 47 in. N/A N/A N/A
19-125-100PM
View Documents
Purus™
PF 2038 01 W 00PM
20 x 38 in. N/A N/A N/A
Description

Description

  • Non-skid backing
  • Protect floors from adhesive residue from the Purus Mats
  • Reduces the risk of slips and falls
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications

Specifications

White
Cleanrooms
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.