Learn More
AG Scientific Inc Puromycin, 1G
Supplier:  AG Scientific Inc P10331GM
Encompass_Preferred
Puromycin Dihydrochloride is an aminoglycoside antibiotic that is isolated from Streptomyces alboniger. Puromycin acts as a protein synthesis inhibitor by incorporating itself into a polypetide chain that has been growing, causing it to prematurely terminate. Puromycin's primary application is as a gene-selection antibiotic with an important role in gene-editing via CRISPR/Cas9.This puromycin reagent, which is in the form of hydrochloride salt, has antineoplastic, antitrypanosomal, and antimicrobial properties. It is important to keep Puromycin Dihydrochloride stored in a tightly closed container in a well-ventilated and dry area.CAS Number: 58-58-2Molecular Weight: 544.43 DaChemical Formula: C22H29N7O5 2HClSolubility: Soluble in water (50 mg/mL), ethanol (~1 mg/mL), DMSO (~13 mg/mL), DMF (~14 mg/mL), and PBS, pH 72 (~10 mg/mL)Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6944
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.