Learn More
Chemglass Life Sciences PTFE Diaphragm Liquid Transfer Pump
SDP
Supplier:  Chemglass Life Sciences CG117120
Encompass_Preferred
• Adjustable Flow up to 3L/min • Self-Priming Up to 13 feet Of Water • No Metal Wetted Parts • Runs Dry Without Damage • Splash Proof Case • Maintenance-Free DesignLiquid transfer pump is excellent for use with corrosive or neutral liquids. Pump is self priming, maintenance-free and will operate dry indefinitely without damage.
Catalog No. 50-401-776
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.