Learn More
Eurofins Discoverx PTH(1-34), 500 µg. (92-1082)
Supplier:  Eurofins Discoverx 921082
Encompass_Preferred
Parathyroid hormone (PTH) is a polypeptide hormone secreted by the parathyroid that enhances bone calcium mobilization, enhances osteoblast activity (anabolic), increases blood calcium levels and calcium:phosphate ratio in the kidney, and promotes absorption of calcium by the intestines. The 1-34 amino acid fragment also induces BMP gene transcription. Agonist of the parathyroid hormone receptors PTH1 and PTH2. Provided as 500 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9621
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.