Learn More
Chemglass Life Sciences FEP Tubing; 1/4" OD x 1/8" ID x 20ft Long
SDP
Supplier:  Chemglass Life Sciences CG197502
Encompass_Preferred
FEP Tubing; 1/4" OD x 1/8" ID x 20ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.
Catalog No. CG197502
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.