Learn More
Andwin Scientific PTFE TAPE 1IN X260IN 12CS
Supplier: Andwin Scientific 44077
The LA-CO Pipe Thread Tape is constructed with PTFE that provides efficient sealing power. This tape can seal on metals and stainless steel within the temperature range of -450�F to 550�F (-268�C to 288�C) and pressure of up to 10,000 PSI (690 BAR). It is non-toxic. The tape measures 1" x 260". It is sold in a case of 12.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.