Learn More
Cytiva Pseudomonas Broth, Pseudomonas sp., 2 mL plastic (50/pkg)
Supplier: Cytiva 4306N
Sterile Ampoule Media Pre-Measured for Maximum Efficiency and Convenience-Meets Membrane Filter (MF) Technique Standard Method requirements with 2 mL ampoules*-Maximizes efficiency with premixed and presterilized media-Choose between plastic or glass ampoules for your applications-Simplify pouring of the media with wide-mouth glass ampoules-All plastic ampoule media is economically packaged with 50 ampoules per box This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.