Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROTOCOL™ Zinc Formalin, Fisher Healthcare™
- Enhances the quality of immunochemical staining
- Minimizes antigen crosslinking
$91.00 - $306.00
Specifications
Chemical Name or Material | PROTOCOL™ Zinc Formalin |
---|---|
Synonym | dihydrogen oxide, dihydrogen monoxide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title