Learn More
Enzo Life Sciences PROTEOSTAT Protein aggregation standards (2x96wells)
Supplier: Enzo Life Sciences ENZ51039KP002
The only commercially available protein aggregation standards assay!. Applications: Fluorescent detection, Microplate. Contents: A. PROTEOSTAT Control (0.0% aggregate). B. PROTEOSTAT Control (0.20% aggregate). C. PROTEOSTAT Control (0.39% aggregate). D. PROTEOSTAT Control (0.78% aggregate). E. PROTEOSTAT Control (1.56% aggregate). F. PROTEOSTAT Control (3.13% aggregate). G. PROTEOSTAT Control (6.25% aggregate). H. PROTEOSTAT Control (12.5% aggregate). Deionized Water, 5 ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.