Learn More
Enzo Life Sciences Protease Inhibitor CocktaIL-(1Set)
SDP
Supplier:  Enzo Life Sciences BMLKI1030001
Encompass_Preferred
Solubility: Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 7X stock solutions in 1.5ml water or 100mM phosphate buffer, pH 7.0. Long Term Storage: +4°C.
Catalog No. 50-201-1086
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.