Learn More
Electron Microscopy Sciences Premium Plus 100-Place Slidebox, Blue
Supplier:  Electron Microscopy Sciences 7149806
Encompass_Preferred
These slideboxes feature a slide rail design that ensures a secure fit with easy placement and removal of slides. Centrally positioned location numbering grid enables simple slide organization with easy viewing of location numbers. They are made from ABS and Polypropylene, which makes the boxes robust and eliminates potential rusting of metal parts. Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement. The slide lock has easily identified lock and unlock symbols and provides secure storage. A 2 mm zip tie can be used for more secure storage, if desired. Holds 100 26 x 75 mm slides.
Color: Blue
Catalog No. 50-192-9962
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.