Learn More
Enzo Life Sciences Pristimerin (5mg). CAS: 1258-84-0
SDP
Supplier:  Enzo Life Sciences ALX350411M005
Encompass_Preferred
Triterpenoid. Antitumor agent. Induces caspase-dependent apoptosis by inhibiting the chymotrypsin-like activity of the proteasome. Inhibits NF-κB activation via inhibition of IKKα or IKKβ. Inhibits induction of inducible nitric oxide synthase (iNOS; NOS II). Shows in vitro antimalarial and insecticidal activity. Induces mitochondrial cell death. Has antiproliferative, antifungal and antiviral properties. Potent and reversible monoacylglycerol lipase (MGL) inhibitor. Alternative name: Celastrol methyl ester. Purity: ≥97% (HPLC). Appearance: Orange crystalline solid. Solubility: Soluble in DMSO or ethyl acetate; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8882
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.