Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kishigo Premium Black Series™ Soft Shell Jacket

Catalog No. 19220978
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Made with high quality materials to increase durability. ML Kishigo™ Premium Black Series™ Soft Shell Jacket gives enhanced visibility with reflective trim and piping.

  • Durable water-resistant soft shell material
  • 2 in. wide high performance reflective material
  • Cuffs feature durable rubber pull tabs for easy adjustments
  • Reflective piping accents add better visibility in low light
  • Convenient underarm vents for breathability
  • Waist cinches hidden in front pockets
  • Black areas offer contrast, style and durability
Pockets: 

  • Two outside lower slash pockets with zipper
  • Left chest media-compatible pocket
  • Left arm cell phone pocket

Compliance

Meets ANSI™ /ISEA 107-2015, Type R, Class 3 standards.

Specifications

Size X-Large
Gender Unisex
Material Polyester
Color Lime
Closure Type Zipper
Certifications/Compliance ANSI/ISEA 107-2015, Type R, Class 3
No. of Pockets 4
Product Type Soft Shell Jacket
Tape Type 2 in. reflective
Thickness 300 gsm
Width (English) 19 in.
Width (Metric) 483 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.