Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Green, 9mm Thread, PTFE/Silicone Septa
SDP
Supplier:  Chemglass Life Sciences CV3800G009
Encompass_Preferred
Preassembled Closure and Septa, Green, 9mm Thread, PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-231-083
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.