Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Black, GPI 13-425, .060" Red PTFE/Silicone Septa with Slit
SDP
Supplier:  Chemglass Life Sciences CV51950013
Encompass_Preferred
Preassembled Closure and Septa, Black, GPI 13-425, .060" Red PTFE/Silicone Septa with Slit. Pre-assembled closure and septa minimize contamination from handling.
Catalog No. 50-231-263
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.