Learn More
Clontech Labs 3P pPUR Vector | 25ug
SDP
Supplier:  Clontech Labs 3P 631601
Encompass
pPUR confers puromycin resistance to mammalian cells and can be used to select for stably transformed cell lines.
Catalog No. NC0775858
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.