Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Safety PowerForm PF-90BK 5 mil Biodegradable Nitrile Gloves with EcoTek
SDP

Catalog No. 17441084
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Biodegradable, single-use gloves combine performance, durability, and tactile sensitivity for critical tasks. Fully textured surface enhances grip capabilities in wet and dry conditions.

  • EcoTek Biodegradability Technology accelerates landfill breakdown without any performance loss
  • Fully textured to enhance grip in wet and dry conditions
  • Clean processing technology reduces potential skin irritation
  • Silicone-free for critical environments
  • Optimal thickness for durability and tactile sensitivity
  • Tear-resistant beaded cuff eases donning
  • Consistent thickness for greater dexterity and tactile sensitivity
  • 1.5 AQL exceeds laboratory standards

Specifications

Certifications/Compliance FDA 21 CFR 177.2600, ASTM D6319-19, ASTM F1671-13, ASTM F739-12
Certifications/Compliance (Europe) EN ISO 21420:2020, EN 455-2000-2015, Reach EU 1907/2006, EU 2016/425
Chemotherapeutic Rated No
Applications Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 5.6 mil
Finger Thickness (Metric) 0.14 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size 2X-Large
Thickness (English) 5 mil
Thickness (Metric) 0.13 mm
Series PF-90BK
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product tested under ASTM D5526-12, a standard test method for determining anaerobic biodegradation of plastics in accelerated landfill conditions. Current biodegradation results do not predict future results.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.