Learn More
CTL Scientific Supply Corp POTASSIUM TEST PAP.,200 STRIPS
SDP
Supplier:  CTL Scientific Supply Corp 90727
Encompass_Preferred
POTASSIUM TEST PAPER - box of 200 strips (each strip 20 x 70 mm). This test paper shows potassium as orange-red spots on a light yellow background. Limit of sensitivity: 250 mg/l K +
Catalog No. 50-117-3295
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.