Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Rhodizonate, MP Biomedicals
Click to view available options
Quantity:
1 g
Specifications
Specifications
Quantity | 1 g |
Formula Weight | 246.26 |
Chemical Name or Material | Potassium Rhodizonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction