Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Potassium Channel Kir3.4, Mouse, Unlabeled, Clone: 8F4.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1721
Description
G protein-activated inward rectifier potassium channel 4 (UniProt P48544; also known as Cardiac inward rectifier, CIR, Heart KATP channel, Inward rectifier K(+) channel Kir3.4, GIRK-4, IRK-4, KATP-1, Potassium channel, inwardly rectifying subfamily J member 5) is encoded by the KCNJ5 (also known as GIRK4, LQT13) gene (Gene ID 3762) in human. Inward rectifier K+ channels (Kirs) modulate cell membrane potential and are classified as K+ transport channels (Kir1.x, Kir4.x, Kir5.x, Kir7.x), classic Kir (Kir2.x), G-protein-gated Kirs (Kir2.x and Kir3.x), and ATP-sensitive Kirs (Kir6.x). Kirs mediate diverse physiological functions of both excitable and nonexcitable epithelial cells, such as cell adhesion-migration, volume regulation, apoptosis, and proliferation. While a large fraction of Kir3.4 (GIRK4; KCNJ5) subunits is processed to the cell membrane to form homomultimers, Kir3.4 subunits also assemble into heteromeric channels with Kir3.1 (GIRK1; KCNJ3) subunits in atrial myocytes or Kir3.2 (GIRK2; KCNJ6) in the cerebellum. Upon agonist activation of appropriate GPCRs, heteroterameric Kir3.4 channels are opened via interaction with subunits of the pertussis toxin-sensitive class (Gi/o) G-proteins, resulting in membrane hyperpolarization and/or reduced excitability. Whereas homotetrameric Kir3.4 channels can be activated by high intracellular Na+ independent of G-protein. Human Kir3.4 is a 419-amino acid multi-pass membrane (a.a. 87-111, 136-147, 164-185) protein that contains two small extraceullar regions (a.a. 112-135 and 155-163) and a pore-forming domain (a.a. 148-154), having both its N- and C-terminal ends exposed intracellularly (a.a. 1-86 and a.a. 186-419).Specifications
Potassium Channel Kir3.4 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
P48544 | |
GST-tagged human potassium channel Kir3.4 C-terminal end fragment. | |
100 μg | |
Signaling | |
NP_000881 | |
Human | |
Purified |
Western Blot | |
8F4.1 | |
Unconjugated | |
KCNJ5, GIRK4, LQT13 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 8F4.1 detects human potassium channel Kir3.4 by targeting an epitope within the C-terminal cytoplasmic domain. | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction