Learn More
Enzo Life Sciences AMPINEXT™ Post-Bisulfite DNA Library Preparation Kit (Illumina) (12Reactions)
Supplier: Enzo Life Sciences ENZGEN5120012
The AMPINEXT™ Post-Bisulfite DNA Library Preparation Kit is a complete set of optimized reagents to prepare a DNA library -- after successful bisulfite conversion -- for various Illumina platform-based bisulfite sequencing (bisulfite-seq) assays, such as whole genome bisulfite sequencing (WGBS), oxidative bisulfite sequencing (oxBs-seq), reduced representation bisulfite sequencing (RRBS), and other bisulfite-based next generation sequencing applications. The optimized protocol and components of the kit allow both non-barcoded (singleplexed) and barcoded (multiplexed) DNA libraries to be quickly constructed using sub-nanogram input concentrations of DNA since the DNA is first bisulfite-converted and then used for library preparation. Assay Time: 5 hours.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.