Learn More
Andwin Scientific POLYSTYRENE RESERVOIR, 25 ML
SDP
Supplier:  Andwin Scientific 89094662
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 25 mL and has a dimension of 14.6 x 5.1 x 2.5 cm. This product is sold in a case of 200 white sterile reservoirs.
Catalog No. NC0775278
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.