Learn More
Enzo Life Sciences Polymyxin B . sulfate (1g). CAS: 1405-20-5
SDP
Supplier:  Enzo Life Sciences ALX380040G001
Encompass_Preferred
Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase. Purity: ≥80% (HPLC) (sum of polymyxins B1, B2, B3, B1-I). Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8925
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.