Learn More
PTS Diagnostics CardioChek™ PA Analyzer
Cost-efficient, user-friendly diagnostic point-of-care analyzer, which can accurately determine multiple patient values
$1111.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
For Use With (Application) | Easy to use, portable, cost- efficient, runs single and multiple analyte panels |
Quantity | Each |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
21-124-376
|
PTS Diagnostics
1708 |
PA Analyzer |
Each of 1 for $1,111.00
|
|
|||||
Description
The CardioChek PA is a state-of-the-art, hand-held, battery-operated, software-driven reflectance spectrophotometer. It operates on two AAA batteries. The battery life yields approximately 300 tests.
- Runs single and multiple analyte panels
- Small sample (fingerstick): 15 to 40μL
- Results in just two minutes
- Large digital readout
- Very minimal start up costs
- Can be programmed to display in seven languages: English, Spanish, French, German, Italian, Portuguese and Dutch
- Analyzes up to 4 cholesterol types w/ a single fingerstick of blood
- Uses 1-panel, 2-panel, 3-panel test strips (sold separately)
- LDL can be measured and/or auto-calculated by analyzer
- 15μl blood capillaries for 1-panel
- 30μl for 2-panel
- 40μl for 3-panel (sold separately)
- Recommend using lancets: device/tips or single-use (sold separately)
Order Info
Factory-calibrated and ready to use; easy to ship and store
Certifications
CLIA-waived, FDA-cleared, CRMLN-certified, CE-marked
Compliance
Meets NCEP guidelines for accuracy and precision
Specifications
CE Marked | |
Easy to use, portable, cost- efficient, runs single and multiple analyte panels | |
Each |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.