Learn More
PTS Diagnostics Accessories for CardioChek™ PA Analyzer
Experience increased versatility and usefulness with these accessories
Supplier:  PTS Diagnostics 11641886
 View more versions of this product
Catalog No. 21-124-380
Explore all special offers
Add to Cart
Description
Description
  • CardioChek PA Printer - Sleek, cost-effective design offers increased portability and light weight; cost-effective printer solution that is significantly less expensive to transport and ship from test site to site
  • CardioChek Link - from CardioChek PA device to computer supported on Windows 7, XP, and Vista
  • CardioChek PA Printer Labels - available as 160 per roll
  • Training instructions for Cardiochek analyzer - easy-to-use CD-ROM
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Each
Software CD and Computer Cable
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.