Learn More
Chemglass Life Sciences Poly Crimp Seal, 11mm, Red Cap, PTFE/Silicone/PTFE Septum
SDP
Supplier:  Chemglass Life Sciences CV3940R011
Encompass_Preferred
Poly Crimp Seal, 11mm, Red Cap, PTFE/Silicone/PTFE Septum. A crimp cap constructed of plastic that fits virtually any 11mm crimp sample vial. Use a standard crimping tool to securely crimp the Poly Crimp seal onto any crimp top vial. The Poly Crimp seal also permits an airtight seal by hand without crimping. Although the Poly Crimp seal can work with any crimp top vial, the tighter manufacturing tolerances and quality assurance program with the vials supplied by Chemglass certifies better performance when the cap is used with Chemglass supplied vials. Vials Compatible with Poly Crimp Seals: CV-1108-1232: Vial, 2.0mL, Clear, Snap Seal, 12 x 32mm, 11mm Crimp CV-1168-1232 : Vial, Standard Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1185-1232 : Vial, Large Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1200-1232 : Vial, Snap Ring, 2.0mL, Clear, 12 x 32mm, 11mm Crimp
Catalog No. 50-153-2611
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.