Learn More
Electron Microscopy Sciences Poly-L-Lysine 0.1% Aqueous Solution, 25988-63-0, 100ml
Supplier: Electron Microscopy Sciences 19321B
Molecular Formula C18H36N6O3X2.3BrH, Poly-L-lysine with a molecular weight of >70,000. This solution is useful in promoting cell adhesion to solid substrates and used for nuclear antigen-coated red cells in hymolytic plaque assay. The 0.1% aqueous solution which is micro-filtered and is used to increase tissue adhesion to the glass slide, which is needed for immunohistochemical and immunolabeling techniques, as well as lengthy staining procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.