Learn More
Electron Microscopy Sciences Poly-L-Lysine 0.1% Aqueous Solution 100 ML
Supplier:  Electron Microscopy Sciences 19321A
Encompass_Preferred
Poly-l-lysine solution. Useful for promoting cell adhesion; preparation of polycationic beads, immobilization of membranes; immobilization of plant protoplasts, a prepared 0.1% aqueous solution, micro-filtered, ideal for cell growth.
Catalog No. 50-190-1141
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.