Learn More
Monarch Instrument Pocket Tach PT99, Portable Tachometer
Supplier: Monarch Instrument 6109010
Pocket Tach 99 is a digital, battery-powered portable non-contact optical tachometer (red LED), which operates up to 36 inches from a reflective tape target using an LED light source. The ergonomic design allows safe, direct line-of-sight viewing of both the rotating target and the display at the same time, while providing a no-slip rubber surface for single hand operation. Pocket Tach 99 in the value-leader of the world-class Pocket Tach Series from Monarch. Power: (2) "AA" 1.5 VDC batteries (20 hours)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.