Learn More
Andwin Scientific PNEUMATIC EGG PUNCH
Supplier: Andwin Scientific 099CE90
The Pneumatic Egg Punch is an automatic tool that allows fast opening of embryonated chicken eggs. This product is handheld.