Learn More
Glass Expansion Platinum Sampler with Copper Core
Supplier:  Glass Expansion TG1026APT/CU
Encompass_Preferred
Platinum Sampler Cone with Copper Core (3601289), for Thermo Q/RQ/TQ and X Series ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use a new Graphite Sampler Cone Gasket (TG5001) each time the cone is re-installed, even if it’s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For common applications with <5% acid matrix, non-HF, non-organic, see catalog number 501694614. For lowest Ni background and laser ablation applications, see catalog number TG1021-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2935
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.