Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plasminogen Polyclonal Antibody, HRP, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A81120P
Description
Plasminogen Polyclonal antibody specifically detects Plasminogen in Human samples. It is validated for ELISA, Western BlotSpecifications
Plasminogen | |
Polyclonal | |
HRP | |
PLG | |
Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1 | |
Antigen affinity chromatography | |
RUO | |
5340 | |
4° C | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.1M NaCl and 0.01% thimerosal; pH 7.4 | |
P00747 | |
Goat | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction