Learn More
Andwin Scientific PIPET-LITE XLS PIPET 20-200UL
Supplier: Andwin Scientific L200XLS
The Pipet-Lite XLS+ manual single-channel pipette features LTS LiteTouch tips that offer accurate, precise, and consistent sample pickup. The pipette also provide comfort with lower spring forces for users. With a secure volume lock, the pipette reduces the risk of accidental drifts and permits adjustment with one hand. This product has a volume range of 20 L 200 L with a manual increment of 0.2 L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.