Learn More
Innovative Research Inc Pig tPA Rcmb Act 100ug
Supplier:  Innovative Research Inc IPCTPAAR100UG
Encompass_Preferred
Porcine tPA Recombinant Active 100ug - Porcine tPA Recombinant Active from Innovative Research has been recombinantly produced in insect cell culture and purified by affinity chromatography. This is a Frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of >95% as determined by SDS-PAGE. More Details Species Porcine Target tPA Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source Insect cell culture Storage Conditions -70 C Additional Information Greater than 95% active. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including cell signaling, clotting assays, fibrinolysis, hemostasis, and more.
Catalog No. 50-274-4117
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.