Learn More
Electron Microscopy Sciences Picric Acid Saturated, Aqueous, 88-89-1, 500ml
Supplier:  Electron Microscopy Sciences 19552
Encompass_Preferred
Molecular Formula C6H3N3O7, Molecular Weight 229.104, Picric Acid (Saturated, Aqueous) is used as solution for fungal testing.
Catalog No. 50-320-49
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.