Learn More
Hach Company Phthalate-Phosphate Reagent Powder Pillows, 25 mL, pk/50
Supplier:  Hach Company 2150166
Encompass_Preferred
For low-level determination of nickel and cobalt by the PAN method. Hach method 8078. Sample Size: 25 mL. Pack of 50 Powder Pillows.
Catalog No. 50-199-5473
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.